accessibility menu, dialog, popup

UserName

CAS RN 7356-11-8

IUPAC Name: methyl 4-methyl-3-nitrobenzoate
Synonyms: methyl 4-methyl-3-nitrobenzoate
Molecular Weight (g/mol): 195.17
Molecular Formula: C9H9NO4
InChi Key: YFPBHPCMYFCRKS-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C)C(=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results