Learn More
Methyl 4-methoxyindole-2-carboxylate, 99%
CAS: 111258-23-2 | C11H11NO3 | 205.21 g/mol
Supplier: Thermo Scientific Chemicals H2774303
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Methyl 4-methoxyindole-2-carboxylate | |
| 140°C to 142°C | |
| MFCD00134300 | |
| methyl 4-methoxy-2-indolecarboxylate, 4-methoxy-1h-indole-2-carboxylic acid methyl ester, methyl 4-methoxyindole-2-carboxylate, 1h-indole-2-carboxylic acid, 4-methoxy-, methyl ester, 4-methoxyindole-2-carboxylic acid methyl ester, methyl-4-methoxy-2-indolecarboxylate, 1h-indole-2-carboxylicacid, 4-methoxy-, methyl ester, methyl4-methoxy-2-indolecarboxylate, pubchem7250, acmc-1btz2 | |
| COC(=O)C1=CC2=C(OC)C=CC=C2N1 | |
| 205.21 | |
| 205.21 |
| 111258-23-2 | |
| C11H11NO3 | |
| 1 g | |
| GLCZQTLCVLVFGV-UHFFFAOYSA-N | |
| methyl 4-methoxy-1H-indole-2-carboxylate | |
| 688172 | |
| 99% |
Chemical Identifiers
| 111258-23-2 | |
| 205.21 | |
| GLCZQTLCVLVFGV-UHFFFAOYSA-N | |
| 688172 | |
| COC(=O)C1=CC2=C(OC)C=CC=C2N1 |
| C11H11NO3 | |
| MFCD00134300 | |
| methyl 4-methoxy-2-indolecarboxylate, 4-methoxy-1h-indole-2-carboxylic acid methyl ester, methyl 4-methoxyindole-2-carboxylate, 1h-indole-2-carboxylic acid, 4-methoxy-, methyl ester, 4-methoxyindole-2-carboxylic acid methyl ester, methyl-4-methoxy-2-indolecarboxylate, 1h-indole-2-carboxylicacid, 4-methoxy-, methyl ester, methyl4-methoxy-2-indolecarboxylate, pubchem7250, acmc-1btz2 | |
| methyl 4-methoxy-1H-indole-2-carboxylate |
Safety and Handling
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only