missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-methoxyindole-2-carboxylate, 99%

Catalog No. p-7030193
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH27743MD 250 mg
AAH2774303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH27743MD Supplier Thermo Scientific Chemicals Supplier No. H27743MD
Only null left

CAS: 111258-23-2 | C11H11NO3 | 205.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 111258-23-2
Molecular Formula C11H11NO3
Molecular Weight (g/mol) 205.21
MDL Number MFCD00134300
InChI Key GLCZQTLCVLVFGV-UHFFFAOYSA-N
Synonym methyl 4-methoxy-2-indolecarboxylate, 4-methoxy-1h-indole-2-carboxylic acid methyl ester, methyl 4-methoxyindole-2-carboxylate, 1h-indole-2-carboxylic acid, 4-methoxy-, methyl ester, 4-methoxyindole-2-carboxylic acid methyl ester, methyl-4-methoxy-2-indolecarboxylate, 1h-indole-2-carboxylicacid, 4-methoxy-, methyl ester, methyl4-methoxy-2-indolecarboxylate, pubchem7250, acmc-1btz2
PubChem CID 688172
IUPAC Name methyl 4-methoxy-1H-indole-2-carboxylate
SMILES COC(=O)C1=CC2=C(OC)C=CC=C2N1

Specifications

Melting Point 140°C to 142°C
Quantity 250 mg
Formula Weight 205.21
Percent Purity 99%
Chemical Name or Material Methyl 4-methoxyindole-2-carboxylate
Hazard Category H315-H319
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only