CAS RN 111258-23-2
IUPAC Name:
methyl 4-methoxy-1H-indole-2-carboxylate
Synonyms:
methyl 4-methoxy-1H-indole-2-carboxylate
Molecular Weight (g/mol):
205.21
Molecular Formula:
C11H11NO3
InChi Key:
GLCZQTLCVLVFGV-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC2=C(OC)C=CC=C2N1
1
–
1
of
1
results
1
Promotions Available
Methyl 4-methoxyindole-2-carboxylate, 99%
Methyl 4-methoxyindole-2-carboxylate, 99%
CAS: 111258-23-2 Molecular Formula: C11H11NO3 Molecular Weight (g/mol): 205.21 MDL Number: MFCD00134300 InChI Key: GLCZQTLCVLVFGV-UHFFFAOYSA-N Synonym: methyl 4-methoxy-2-indolecarboxylate,4-methoxy-1h-indole-2-carboxylic acid methyl ester,methyl 4-methoxyindole-2-carboxylate,1h-indole-2-carboxylic acid, 4-methoxy-, methyl ester,4-methoxyindole-2-carboxylic acid methyl ester,methyl-4-methoxy-2-indolecarboxylate,1h-indole-2-carboxylicacid, 4-methoxy-, methyl ester,methyl4-methoxy-2-indolecarboxylate,pubchem7250,acmc-1btz2 PubChem CID: 688172 IUPAC Name: methyl 4-methoxy-1H-indole-2-carboxylate SMILES: COC(=O)C1=CC2=C(OC)C=CC=C2N1