Learn More
Methyl 4-methoxybenzoate, 98+%
CAS: 121-98-2 | C9H10O3 | 166.176 g/mol
Supplier: Thermo Scientific Chemicals A1259814
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Methyl 4-methoxybenzoate | |
| 48°C to 51°C | |
| >110°C (230°F) | |
| C9H10O3 | |
| 25 g | |
| methyl anisate, methyl p-anisate, methyl p-methoxybenzoate, 4-methoxybenzoic acid methyl ester, p-anisic acid, methyl ester, benzoic acid, 4-methoxy-, methyl ester, methyl 4-anisate, p-methoxybenzoic acid methyl ester, p-anisic acid methyl ester, benzoic acid, p-methoxy-, methyl ester | |
| COC1=CC=C(C=C1)C(=O)OC | |
| 166.176 | |
| CHEBI:86903 | |
| ≥98% |
| 121-98-2 | |
| 244°C to 245°C | |
| Characteristic | |
| MFCD00008437 | |
| 2208571 | |
| DDIZAANNODHTRB-UHFFFAOYSA-N | |
| methyl 4-methoxybenzoate | |
| 8499 | |
| 166.18 |
Chemical Identifiers
| 121-98-2 | |
| 166.176 | |
| DDIZAANNODHTRB-UHFFFAOYSA-N | |
| 8499 | |
| methyl 4-methoxybenzoate |
| C9H10O3 | |
| MFCD00008437 | |
| methyl anisate, methyl p-anisate, methyl p-methoxybenzoate, 4-methoxybenzoic acid methyl ester, p-anisic acid, methyl ester, benzoic acid, 4-methoxy-, methyl ester, methyl 4-anisate, p-methoxybenzoic acid methyl ester, p-anisic acid methyl ester, benzoic acid, p-methoxy-, methyl ester | |
| CHEBI:86903 | |
| COC1=CC=C(C=C1)C(=O)OC |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 204-513-2
RTECSNumber : BZ4925000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only