missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-methoxybenzoate, 98+%

Catalog No. p-7020588
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1259814 25 g
AAA1259822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1259814 Supplier Thermo Scientific Chemicals Supplier No. A1259814

CAS: 121-98-2 | C9H10O3 | 166.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 121-98-2
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00008437
InChI Key DDIZAANNODHTRB-UHFFFAOYSA-N
Synonym methyl anisate, methyl p-anisate, methyl p-methoxybenzoate, 4-methoxybenzoic acid methyl ester, p-anisic acid, methyl ester, benzoic acid, 4-methoxy-, methyl ester, methyl 4-anisate, p-methoxybenzoic acid methyl ester, p-anisic acid methyl ester, benzoic acid, p-methoxy-, methyl ester
PubChem CID 8499
ChEBI CHEBI:86903
IUPAC Name methyl 4-methoxybenzoate
SMILES COC1=CC=C(C=C1)C(=O)OC

Specifications

Melting Point 48°C to 51°C
Boiling Point 244°C to 245°C
Flash Point >110°C (230°F)
Odor Characteristic
Quantity 25 g
Beilstein 2208571
Formula Weight 166.18
Percent Purity ≥98%
Chemical Name or Material Methyl 4-methoxybenzoate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 204-513-2
RTECSNumber BZ4925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only