missing translation for 'onlineSavingsMsg'
Learn More

Méthyl 4-Méthoxy-3-nitrobenzoate 98,0+%, TCI America™

Numéro de catalogue. M28735G
Modifier l'affichage
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
M28735G 5 g
M28731G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. M28735G Fournisseur TCI America Code fournisseur M28735G

Identifiants chimiques

CAS 40757-20-8
Molecular Formula C9H9NO5
Molecular Weight (g/mol) 211.17
MDL Number MFCD00024213
InChI Key ZUZYMTBOKNSYEB-UHFFFAOYSA-N
Synonym benzoic acid, 4-methoxy-3-nitro-, methyl ester, 3-nitro-4-methoxybenzoic acid methyl ester, methyl 3-nitro-p-anisate, acmc-1ap03, methyl 3-nitro-4-methoxybenzoate, methyl-4-methoxy-3-nitrobenzoate, 3-nitro-p-anisic acid methyl ester, 4-methoxy-3-nitrobenzoic acid methyl ester, 4-methoxy-3-nitro-benzoic acid methyl ester, benzoic acid, 4-methoxy-5-nitro-, methyl ester
PubChem CID 602104
IUPAC Name méthyle 4-méthoxy-3-nitrobenzoate
SMILES COC(=O)C1=CC=C(OC)C(=C1)[N+]([O-])=O

Spécifications

Melting Point 110°C
Color Jaune
Quantity 5 g
Formula Weight 211.17
Percent Purity ≥98.0% (GC)
Physical Form Poudre cristalline
Chemical Name or Material Methyl 4-Methoxy-3-nitrobenzoate
EINECSNumber (3)-1599&(3)-3058
TSCA No