accessibility menu, dialog, popup

UserName

CAS RN 40757-20-8

IUPAC Name: methyl 4-methoxy-3-nitrobenzoate
Synonyms: methyl 4-methoxy-3-nitrobenzoate
Molecular Weight (g/mol): 211.17
Molecular Formula: C9H9NO5
InChi Key: ZUZYMTBOKNSYEB-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(OC)C(=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results