missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Methoxy-3-nitrobenzoate 98.0+%, TCI America™

Catalog No. M28731G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28731G 1 g
M28735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28731G Supplier TCI America Supplier No. M28731G

Chemical Identifiers

CAS 40757-20-8
Molecular Formula C9H9NO5
Molecular Weight (g/mol) 211.17
MDL Number MFCD00024213
InChI Key ZUZYMTBOKNSYEB-UHFFFAOYSA-N
Synonym benzoic acid, 4-methoxy-3-nitro-, methyl ester, 3-nitro-4-methoxybenzoic acid methyl ester, methyl 3-nitro-p-anisate, acmc-1ap03, methyl 3-nitro-4-methoxybenzoate, methyl-4-methoxy-3-nitrobenzoate, 3-nitro-p-anisic acid methyl ester, 4-methoxy-3-nitrobenzoic acid methyl ester, 4-methoxy-3-nitro-benzoic acid methyl ester, benzoic acid, 4-methoxy-5-nitro-, methyl ester
PubChem CID 602104
IUPAC Name methyl 4-methoxy-3-nitrobenzoate
SMILES COC(=O)C1=CC=C(OC)C(=C1)[N+]([O-])=O

Specifications

Melting Point 110°C
Color Yellow
Quantity 1 g
Formula Weight 211.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Methoxy-3-nitrobenzoate
EINECSNumber (3)-1599&(3)-3058
TSCA No