missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Iodo-3-methylbenzoate 98.0+%, TCI America™

Catalog No. I06015G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I06015G 5 g
I060125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I06015G Supplier TCI America Supplier No. I06015G

Chemical Identifiers

CAS 5471-81-8
Molecular Formula C9H9IO2
Molecular Weight (g/mol) 276.073
MDL Number MFCD02683863
InChI Key HCSGWQGKCVQIRM-UHFFFAOYSA-N
Synonym 4-iodo-3-methylbenzoic acid methyl ester, methyl 4-iodo-3-methyl-benzoate, 4-iodo-m-toluic acid methyl ester, benzoic acid, 4-iodo-3-methyl-, methyl ester, methyl4-iodo-3-methylbenzoate, acmc-1anqa, methyl 4-iodo-m-toluate, methyl 3methyl-4-iodobenzoate, methyl 3-methyl-4-iodobenzoate, 4-iodo-m-toluic acid, methyl ester
PubChem CID 231757
IUPAC Name methyl 4-iodo-3-methylbenzoate
SMILES CC1=C(C=CC(=C1)C(=O)OC)I

Specifications

Melting Point 61°C
Boiling Point 106°C
Quantity 5 g
Formula Weight 276.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Iodo-3-methylbenzoate