accessibility menu, dialog, popup

UserName

CH3OOIH3C

CAS RN 5471-81-8

IUPAC Name: methyl 4-iodo-3-methylbenzoate
Synonyms: methyl 4-iodo-3-methylbenzoate
Molecular Weight (g/mol): 276.07
Molecular Formula: C9H9IO2
InChi Key: HCSGWQGKCVQIRM-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(I)C(C)=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)