missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-hydroxy-3-nitrobenzoate, 98+%

Catalog No. AAA1088018
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1088018 50 g
AAA1088030 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1088018 Supplier Thermo Scientific Chemicals Supplier No. A1088018
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 99-42-3 | C8H7NO5 | 197.146 g/mol

4-alkoxy-3-nitrobenzoic acids were obtained by the alkylation of methyl 4-hydroxy-3-nitrobenzoate with corresponding n-alkylbromides in 3-pentanone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-alkoxy-3-nitrobenzoic acids were obtained by the alkylation of methyl 4-hydroxy-3-nitrobenzoate with corresponding n-alkylbromides in 3-pentanone.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 99-42-3
Molecular Formula C8H7NO5
Molecular Weight (g/mol) 197.146
MDL Number MFCD00014695
InChI Key GNCWCTBHZCBXGL-UHFFFAOYSA-N
Synonym methyl 3-nitro-4-hydroxybenzoate, benzoic acid, 4-hydroxy-3-nitro-, methyl ester, 4-hydroxy-3-nitrobenzoic acid methyl ester, methyl 3-nitro-4-hydroxy toluate, aurora 6897, 3-nitro-4-hydroxybenzoic acid methyl ester, methyl-4-hydroxy-3-nitrobenzoic acid, 3-nitro-4-hydroxymethylbenzoate, acmc-209scn, 3-10-00-00378 beilstein handbook reference
PubChem CID 66835
IUPAC Name methyl 4-hydroxy-3-nitrobenzoate
SMILES COC(=O)C1=CC(=C(C=C1)O)[N+](=O)[O-]

Specifications

Melting Point 73°C to 77°C
Quantity 50 g
Beilstein 2115197
Formula Weight 197.15
Percent Purity ≥98%
Chemical Name or Material Methyl 4-hydroxy-3-nitrobenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 202-755-3
RTECSNumber DH2528400
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only