accessibility menu, dialog, popup

UserName

CAS RN 99-42-3

IUPAC Name: methyl 4-hydroxy-3-nitrobenzoate
Synonyms: methyl 4-hydroxy-3-nitrobenzoate
Molecular Weight (g/mol): 197.15
Molecular Formula: C8H7NO5
InChi Key: GNCWCTBHZCBXGL-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(O)C(=C1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results