missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-fluorophenylacetate, 98%

Catalog No. AAH3223606
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3223606 5 g
AAH3223614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3223606 Supplier Thermo Scientific Chemicals Supplier No. H3223606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 34837-84-8 | C9H9FO2 | 168.167 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 34837-84-8
Molecular Formula C9H9FO2
Molecular Weight (g/mol) 168.167
MDL Number MFCD00800608
InChI Key AJPPKGMEHMXPMC-UHFFFAOYSA-N
Synonym methyl 2-4-fluorophenyl acetate, methyl 4-fluorophenylacetate, 4-fluorophenylacetic acid methyl ester, pubchem24389, acmc-209iaq, methyl-4-fluorphenylacetate, methyl-4-fluorophenylacetate, methyl 4-fluorophenyl-acetate, methyl 4-fluorophenyl acetate
PubChem CID 2733233
IUPAC Name methyl 2-(4-fluorophenyl)acetate
SMILES COC(=O)CC1=CC=C(C=C1)F

Specifications

Quantity 5 g
Formula Weight 168.17
Percent Purity 98%
Chemical Name or Material Methyl 4-fluorophenylacetate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only