missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Fluorobenzoate 97.0+%, TCI America™

Catalog No. p-7129666
Change view
Click to view available options
Quantity:
25 g
500 g
Catalog No. Quantity
F040325G 25 g
F0403500G 500 g
2 options

Catalog No. F040325G

Supplier: TCI America F040325G

Only null left

Chemical Identifiers

CAS 403-33-8
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00017959
InChI Key MSEBQGULDWDIRW-UHFFFAOYSA-N
Synonym methyl p-fluorobenzoate, 4-fluorobenzoic acid methyl ester, methyl 4-fluorobenzoic acid, benzoic acid, 4-fluoro-, methyl ester, methyl4-fluorobenzoate, methyl-4-fluorobenzoate, 4-fluoro-benzoic acid methyl ester, pubchem3566, acmc-1aef0, asischem p32019
PubChem CID 67878
IUPAC Name methyl 4-fluorobenzoate
SMILES COC(=O)C1=CC=C(C=C1)F

Specifications

Color Yellow
Boiling Point 197°C
Quantity 25 g
Formula Weight 154.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl 4-Fluorobenzoate