accessibility menu, dialog, popup

UserName

CH3OOF

CAS RN 403-33-8

IUPAC Name: methyl 4-fluorobenzoate
Synonyms: methyl 4-fluorobenzoate
Molecular Weight (g/mol): 154.14
Molecular Formula: C8H7FO2
InChi Key: MSEBQGULDWDIRW-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(F)C=C1
Boiling Point 
  • (3)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)