missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-fluorobenzoate, 97%

Catalog No. p-7025817
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Catalog No. Quantity
AAB2150406 5 g
AAB2150414 25 g
AAB2150422 100 g
3 options

Catalog No. AAB2150406

Supplier: Thermo Scientific Chemicals B2150406

Only null left

CAS: 403-33-8 | C8H7FO2 | 154.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 403-33-8
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00017959
InChI Key MSEBQGULDWDIRW-UHFFFAOYSA-N
Synonym methyl p-fluorobenzoate, 4-fluorobenzoic acid methyl ester, methyl 4-fluorobenzoic acid, benzoic acid, 4-fluoro-, methyl ester, methyl4-fluorobenzoate, methyl-4-fluorobenzoate, 4-fluoro-benzoic acid methyl ester, pubchem3566, acmc-1aef0, asischem p32019
PubChem CID 67878
IUPAC Name methyl 4-fluorobenzoate
SMILES COC(=O)C1=CC=C(C=C1)F

Specifications

Density 1.192
Boiling Point 195°C to 197°C
Flash Point 77°C (170°F)
Odor Aromatic
Refractive Index 1.495
Quantity 5 g
Beilstein 2085925
Formula Weight 154.14
Percent Purity 97%
Chemical Name or Material Methyl 4-fluorobenzoate
Hazard Category H227-H302-H315-H318-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P370+P378q-P501c
EINECSNumber 206-956-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only