missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Fluoro-3-hydroxybenzoate 98.0+%, TCI America™

Catalog No. M19195G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19195G 5 g
M191925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M19195G Supplier TCI America Supplier No. M19195G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 214822-96-5
Molecular Formula C8H7FO3
Molecular Weight (g/mol) 170.139
MDL Number MFCD06797457
InChI Key CUGWNEOTLGLGDG-UHFFFAOYSA-N
PubChem CID 12016892
IUPAC Name methyl 4-fluoro-3-hydroxybenzoate
SMILES COC(=O)C1=CC(=C(C=C1)F)O

Specifications

Melting Point 92°C
Color White-Yellow
Quantity 5 g
Synonym 4-fluoro-3-hydroxybenzoic acid methyl ester, benzoic acid, 4-fluoro-3-hydroxy-, methyl ester, pubchem3567, acmc-209fky, methyl4-fluoro-3-hydroxybenzoate, methyl 4-fluoranyl-3-oxidanyl-benzoate, 3-hydroxy-4-fluorobenzoic acid methyl ester, 4-fluoro-3-hydroxy-benzoic acid methyl ester, benzoic acid,4-fluoro-3-hydroxy-, methyl ester
Formula Weight 170.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Fluoro-3-hydroxybenzoate