accessibility menu, dialog, popup

UserName

CAS RN 214822-96-5

IUPAC Name: methyl 4-fluoro-3-hydroxybenzoate
Synonyms: methyl 4-fluoro-3-hydroxybenzoate
Molecular Weight (g/mol): 170.14
Molecular Formula: C8H7FO3
InChi Key: CUGWNEOTLGLGDG-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(O)=C(F)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)