missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-fluoro-2-methylbenzoate, 98%

Catalog No. AAH6326806
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6326806 5 g
AAH6326814 25 g
AAH6326822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6326806 Supplier Thermo Scientific Chemicals Supplier No. H6326806

CAS: 174403-69-1 | C9H9FO2 | 168.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 174403-69-1
Molecular Formula C9H9FO2
Molecular Weight (g/mol) 168.17
MDL Number MFCD06203790
InChI Key ZQFCTCYDRQFPBU-UHFFFAOYSA-N
Synonym benzoic acid, 4-fluoro-2-methyl-, methyl ester, 4-fluoro-2-methylbenzoic acid methyl ester, rarechem al bf 0506, methyl=4-fluoro-2-methylbenzoate, 4-fluoro-2-methyl benzoic acid methyl ester, 4-fluoro-2-methyl-benzoic acid methyl ester, benzoic acid,4-fluoro-2-methyl-,methyl ester
PubChem CID 2782181
IUPAC Name methyl 4-fluoro-2-methylbenzoate
SMILES COC(=O)C1=C(C)C=C(F)C=C1

Specifications

Color Colorless
Quantity 5 g
Formula Weight 168.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methyl 4-fluoro-2-methylbenzoate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only