missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-fluoro-2-methoxybenzoate, 97+%, Thermo Scientific Chemicals

Catalog No. p-7034677
Change view
Click to view available options
Quantity:
5 g
Catalog No. Quantity
AAH5237006 5 g
1 options

Catalog No. AAH5237006

Supplier: Thermo Scientific Chemicals H5237006

Only null left

Methyl 4-fluoro-2-methoxybenzoate  is used as a catalyst, kinase inhibitors and pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4-fluoro-2-methoxybenzoate  is used as a catalyst, kinase inhibitors and pharmaceutical intermediates.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 204707-42-6
Molecular Formula C9H9FO3
Molecular Weight (g/mol) 184.17
MDL Number MFCD06203990
InChI Key LJUAEPNTXDJBRX-UHFFFAOYSA-N
Synonym 4-fluoro-2-methoxybenzoic acid methyl ester, methyl 2-methoxy-4-fluoro-benzoate, benzoic acid, 4-fluoro-2-methoxy-, methyl ester, rarechem al bf 1023, methyl-4-fluoro-2-methoxybenzoate, methyl 4-fluoranyl-2-methoxy-benzoate, 2-methoxy-4-fluorobenzoic acid methyl ester, 4-fluoro-2-methoxy-benzoic acid methyl ester
PubChem CID 10241478
IUPAC Name methyl 4-fluoro-2-methoxybenzoate
SMILES COC(=O)C1=C(OC)C=C(F)C=C1

Specifications

Quantity 5 g
Formula Weight 184.17
Percent Purity ≥97%
Chemical Name or Material Methyl 4-fluoro-2-methoxybenzoate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only