Learn More
Methyl 4-cyanobenzoate, 99%
CAS: 1129-35-7 | C9H7NO2 | 161.16 g/mol
Supplier: Thermo Scientific Chemicals L0836506
Description
It finds its application in the chemoselective biotransformation of nitriles by Rhodococcus equi A4 and in the design of 3-(4-carboxybenzoyl)-2-quinolinecarboxaldehyde as a reagent for ultrasensitive determination of primary amines by capillary electrophoresis using laser fluorescence detection.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt finds its application in the chemoselective biotransformation of nitriles by Rhodococcus equi A4 and in the design of 3-(4-carboxybenzoyl)-2-quinolinecarboxaldehyde as a reagent for ultrasensitive determination of primary amines by capillary electrophoresis using laser fluorescence detection.
Solubility
Insoluble in water. Soluble in chloroform and ethyl acetate.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
| Methyl 4-cyanobenzoate | |
| 65°C to 69°C | |
| C9H7NO2 | |
| 5 g | |
| 4-cyanobenzoic acid methyl ester, benzoic acid, 4-cyano-, methyl ester, methyl p-cyanobenzoate, unii-f8fms7o8xg, f8fms7o8xg, benzoic acid, p-cyano-, methyl ester, 4-cyano-benzoic acid methyl ester, methyl4-cyanobenzoate, methyl-4-cyanobenzoate, acmc-1bs5i | |
| KKZMIDYKRKGJHG-UHFFFAOYSA-N | |
| methyl 4-cyanobenzoate | |
| 70791 | |
| 99% |
| 1129-35-7 | |
| 142°C to 144°C (12 mmHg) | |
| MFCD00001823 | |
| 1866980 | |
| Insoluble in water. Soluble in chloroform and ethyl acetate. | |
| COC(=O)C1=CC=C(C=C1)C#N | |
| 161.16 | |
| 161.16 |
Chemical Identifiers
| 1129-35-7 | |
| 161.16 | |
| KKZMIDYKRKGJHG-UHFFFAOYSA-N | |
| 70791 | |
| COC(=O)C1=CC=C(C=C1)C#N |
| C9H7NO2 | |
| MFCD00001823 | |
| 4-cyanobenzoic acid methyl ester, benzoic acid, 4-cyano-, methyl ester, methyl p-cyanobenzoate, unii-f8fms7o8xg, f8fms7o8xg, benzoic acid, p-cyano-, methyl ester, 4-cyano-benzoic acid methyl ester, methyl4-cyanobenzoate, methyl-4-cyanobenzoate, acmc-1bs5i | |
| methyl 4-cyanobenzoate |
Safety and Handling
GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
EINECSNumber : 214-443-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only