missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-cyanobenzoate, 99%

Catalog No. p-7044816
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0836506 5 g
AAL0836514 25 g
AAL0836522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0836506 Supplier Thermo Scientific Chemicals Supplier No. L0836506

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1129-35-7 | C9H7NO2 | 161.16 g/mol

It finds its application in the chemoselective biotransformation of nitriles by Rhodococcus equi A4 and in the design of 3-(4-carboxybenzoyl)-2-quinolinecarboxaldehyde as a reagent for ultrasensitive determination of primary amines by capillary electrophoresis using laser fluorescence detection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the chemoselective biotransformation of nitriles by Rhodococcus equi A4 and in the design of 3-(4-carboxybenzoyl)-2-quinolinecarboxaldehyde as a reagent for ultrasensitive determination of primary amines by capillary electrophoresis using laser fluorescence detection.

Solubility
Insoluble in water. Soluble in chloroform and ethyl acetate.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 1129-35-7
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00001823
InChI Key KKZMIDYKRKGJHG-UHFFFAOYSA-N
Synonym 4-cyanobenzoic acid methyl ester, benzoic acid, 4-cyano-, methyl ester, methyl p-cyanobenzoate, unii-f8fms7o8xg, f8fms7o8xg, benzoic acid, p-cyano-, methyl ester, 4-cyano-benzoic acid methyl ester, methyl4-cyanobenzoate, methyl-4-cyanobenzoate, acmc-1bs5i
PubChem CID 70791
IUPAC Name methyl 4-cyanobenzoate
SMILES COC(=O)C1=CC=C(C=C1)C#N

Specifications

Melting Point 65°C to 69°C
Boiling Point 142°C to 144°C (12 mmHg)
Quantity 5 g
Beilstein 1866980
Solubility Information Insoluble in water. Soluble in chloroform and ethyl acetate.
Formula Weight 161.16
Percent Purity 99%
Chemical Name or Material Methyl 4-cyanobenzoate
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 214-443-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only