accessibility menu, dialog, popup

UserName

CAS RN 1129-35-7

IUPAC Name: methyl 4-cyanobenzoate
Synonyms: methyl 4-cyanobenzoate
Molecular Weight (g/mol): 161.16
Molecular Formula: C9H7NO2
InChi Key: KKZMIDYKRKGJHG-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C=C1)C#N
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)