missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Chlorobenzoate 98.0+%, TCI America™

Catalog No. C162525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C162525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C162525G Supplier TCI America Supplier No. C162525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1126-46-1
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
MDL Number MFCD00000621
InChI Key LXNFVVDCCWUUKC-UHFFFAOYSA-N
Synonym methyl p-chlorobenzoate, 4-chlorobenzoic acid methyl ester, benzoic acid, 4-chloro-, methyl ester, unii-m0u80j80va, benzoic acid, p-chloro-, methyl ester, methyl-p-chlorobenzoate, p-chlorobenzoic acid methyl ester, methyl-4-chlorobenzoate, 4-chloro-benzoic acid methyl ester, decitabine related compound d
PubChem CID 14307
IUPAC Name methyl 4-chlorobenzoate
SMILES COC(=O)C1=CC=C(Cl)C=C1

Specifications

Melting Point 43°C
Color White-Yellow
Boiling Point 137°C
Quantity 25 g
Formula Weight 170.59
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Chlorobenzoate
TSCA Yes
Recommended Storage Refrigerator