accessibility menu, dialog, popup

UserName

CH3OOCl

CAS RN 1126-46-1

IUPAC Name: methyl 4-chlorobenzoate
Synonyms: methyl P-chlorobenzoate
Molecular Weight (g/mol): 170.59
Molecular Formula: C8H7ClO2
InChi Key: LXNFVVDCCWUUKC-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(Cl)C=C1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)