missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-chlorobenzoate, 99%

Catalog No. p-7080543
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1010914 25 g
AAA1010922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1010914 Supplier Thermo Scientific Chemicals Supplier No. A1010914
Only null left

CAS: 1126-46-1 | C8H7ClO2 | 170.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1126-46-1
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
MDL Number MFCD00000621
InChI Key LXNFVVDCCWUUKC-UHFFFAOYSA-N
Synonym methyl p-chlorobenzoate, 4-chlorobenzoic acid methyl ester, benzoic acid, 4-chloro-, methyl ester, unii-m0u80j80va, benzoic acid, p-chloro-, methyl ester, methyl-p-chlorobenzoate, p-chlorobenzoic acid methyl ester, methyl-4-chlorobenzoate, 4-chloro-benzoic acid methyl ester, decitabine related compound d
PubChem CID 14307
IUPAC Name methyl 4-chlorobenzoate
SMILES COC(=O)C1=CC=C(Cl)C=C1

Specifications

Melting Point 41°C to 45°C
Density 1.382
Boiling Point 136°C to 137°C (10 mmHg)
Flash Point 106°C (222°F)
Quantity 25 g
Beilstein 1564310
Merck Index 14,2125
Formula Weight 170.6
Percent Purity 99%
Chemical Name or Material Methyl 4-chlorobenzoate
EINECSNumber 214-420-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only