missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Chloro-3-nitrobenzoate 98.0+%, TCI America™

Catalog No. p-7132850
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M17795G 5 g
M177925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M17795G Supplier TCI America Supplier No. M17795G

Chemical Identifiers

CAS 14719-83-6
Molecular Formula C8H6ClNO4
Molecular Weight (g/mol) 215.59
MDL Number MFCD00016989
InChI Key XRTKWPWDSUNLHS-UHFFFAOYSA-N
Synonym methyl 4-chloro-3-nitrobenzate, 4-chloro-3-nitrobenzoic acid methyl ester, methyl-4-chloro-3-nitrobenzoate, benzoic acid, 4-chloro-3-nitro-, methyl ester, acmc-209cya, methyl-3-nitro-4-chlorobenzoate, methyl 3-nitro-4-chlorobenzoate, xrtkwpwdsunlhs-uhfffaoysa, methyl 4-chloro-3-nitro-benzoate, methyl4-chloro-3-nitrobenzoate
PubChem CID 735797
IUPAC Name methyl 4-chloro-3-nitrobenzoate
SMILES COC(=O)C1=CC=C(Cl)C(=C1)[N+]([O-])=O

Specifications

Melting Point 82°C
Quantity 5 g
Formula Weight 215.59
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Chloro-3-nitrobenzoate
EINECSNumber (3)-1519
TSCA No