accessibility menu, dialog, popup

UserName

CH3OOClNOO

CAS RN 14719-83-6

IUPAC Name: methyl 4-chloro-3-nitrobenzoate
Synonyms: methyl 4-chloro-3-nitrobenzoate
Molecular Weight (g/mol): 215.59
Molecular Formula: C8H6ClNO4
InChi Key: XRTKWPWDSUNLHS-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(Cl)C(=C1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)