missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-chloro-3-nitrobenzoate, 98%

Catalog No. p-7044611
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0740706 5 g
AAL0740714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0740706 Supplier Thermo Scientific Chemicals Supplier No. L0740706

CAS: 14719-83-6 | C8H6ClNO4 | 215.59 g/mol

Methyl 4-chloro-3-nitrobenzoate, is used as starting material for the preparation of the methyl benzoates

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 4-chloro-3-nitrobenzoate, is used as starting material for the preparation of the methyl benzoates

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 14719-83-6
Molecular Formula C8H6ClNO4
Molecular Weight (g/mol) 215.59
MDL Number MFCD00016989
InChI Key XRTKWPWDSUNLHS-UHFFFAOYSA-N
Synonym methyl 4-chloro-3-nitrobenzate, 4-chloro-3-nitrobenzoic acid methyl ester, methyl-4-chloro-3-nitrobenzoate, benzoic acid, 4-chloro-3-nitro-, methyl ester, acmc-209cya, methyl-3-nitro-4-chlorobenzoate, methyl 3-nitro-4-chlorobenzoate, xrtkwpwdsunlhs-uhfffaoysa, methyl 4-chloro-3-nitro-benzoate, methyl4-chloro-3-nitrobenzoate
PubChem CID 735797
IUPAC Name methyl 4-chloro-3-nitrobenzoate
SMILES COC(=O)C1=CC=C(Cl)C(=C1)[N+]([O-])=O

Specifications

Melting Point 78°C to 82°C
Quantity 5 g
Beilstein 1643729
Solubility Information Soluble in water.
Formula Weight 215.59
Percent Purity 98%
Chemical Name or Material Methyl 4-chloro-3-nitrobenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only