missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 4-(Bromomethyl)-3-methoxybenzoate 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 93°C |
| Color | White-Yellow |
| Quantity | 5 g |
| UN Number | 3077 |
| Formula Weight | 259.10 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Methyl 4-(Bromomethyl)-3-methoxybenzoate |
Safety and Handling
| TSCA | No |