accessibility menu, dialog, popup

UserName

CAS RN 70264-94-7

H3COOOH3CBr

CAS RN 70264-94-7

IUPAC Name: methyl 4-(bromomethyl)-3-methoxybenzoate
Synonyms: methyl 4-(bromomethyl)-3-methoxybenzoate
Molecular Weight (g/mol): 259.1
Molecular Formula: C10H11BrO3
InChi Key: UXSNXOMMJXTFEG-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(OC)=C(CBr)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results