missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 4-Bromocrotonate 80.0+%, TCI America™
$59.69 - $152.08
Chemical Identifiers
| CAS | 1117-71-1 |
|---|---|
| Molecular Formula | C5H7BrO2 |
| Molecular Weight (g/mol) | 179.01 |
| MDL Number | MFCD00000246,MFCD09998308 |
| InChI Key | RWIKCBHOVNDESJ-NSCUHMNNSA-N |
| Synonym | methyl 4-bromocrotonate, e-methyl 4-bromobut-2-enoate, 4-bromocrotonic acid methyl ester, methyl gamma-bromocrotonate, methyl bromocrotonate, methyl 4-bromo-2-butenoate, 2-butenoic acid, 4-bromo-, methyl ester, methyl 4-bromobut-2-enoate, methyl 2e-4-bromobut-2-enoate, crotonic acid, 4-bromo-, methyl ester |
| PubChem CID | 5369175 |
| IUPAC Name | methyl (2E)-4-bromobut-2-enoate |
| SMILES | COC(=O)\C=C\CBr |
Chemical Identifiers
| 1117-71-1 | |
| 179.01 | |
| RWIKCBHOVNDESJ-NSCUHMNNSA-N | |
| 5369175 | |
| COC(=O)\C=C\CBr |
| C5H7BrO2 | |
| MFCD00000246,MFCD09998308 | |
| methyl 4-bromocrotonate, e-methyl 4-bromobut-2-enoate, 4-bromocrotonic acid methyl ester, methyl gamma-bromocrotonate, methyl bromocrotonate, methyl 4-bromo-2-butenoate, 2-butenoic acid, 4-bromo-, methyl ester, methyl 4-bromobut-2-enoate, methyl 2e-4-bromobut-2-enoate, crotonic acid, 4-bromo-, methyl ester | |
| methyl (2E)-4-bromobut-2-enoate |
Specifications
| 1117-71-1 | |
| C5H7BrO2 | |
| 5 g | |
| methyl 4-bromocrotonate, e-methyl 4-bromobut-2-enoate, 4-bromocrotonic acid methyl ester, methyl gamma-bromocrotonate, methyl bromocrotonate, methyl 4-bromo-2-butenoate, 2-butenoic acid, 4-bromo-, methyl ester, methyl 4-bromobut-2-enoate, methyl 2e-4-bromobut-2-enoate, crotonic acid, 4-bromo-, methyl ester | |
| COC(=O)\C=C\CBr | |
| 179.01 | |
| 179.01 | |
| Liquid |
| 75°C | |
| MFCD00000246,MFCD09998308 | |
| 1760 | |
| RWIKCBHOVNDESJ-NSCUHMNNSA-N | |
| methyl (2E)-4-bromobut-2-enoate | |
| 5369175 | |
| ≥80.0% (GC) | |
| Methyl 4-Bromocrotonate |
Safety and Handling
RTECSNumber : GQ3120000
TSCA : Yes
Recommended Storage : Refrigerator