missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 4-Bromocrotonate 80.0+%, TCI America™
Supplier: TCI America M168925G
Specifications
| Methyl 4-Bromocrotonate | |
| 75°C | |
| MFCD00000246,MFCD09998308 | |
| 1760 | |
| RWIKCBHOVNDESJ-NSCUHMNNSA-N | |
| methyl (2E)-4-bromobut-2-enoate | |
| 5369175 | |
| ≥80.0% (GC) |
| 1117-71-1 | |
| C5H7BrO2 | |
| 25 g | |
| methyl 4-bromocrotonate, e-methyl 4-bromobut-2-enoate, 4-bromocrotonic acid methyl ester, methyl gamma-bromocrotonate, methyl bromocrotonate, methyl 4-bromo-2-butenoate, 2-butenoic acid, 4-bromo-, methyl ester, methyl 4-bromobut-2-enoate, methyl 2e-4-bromobut-2-enoate, crotonic acid, 4-bromo-, methyl ester | |
| COC(=O)\C=C\CBr | |
| 179.01 | |
| 179.01 | |
| Liquid |
Chemical Identifiers
| 1117-71-1 | |
| 179.01 | |
| RWIKCBHOVNDESJ-NSCUHMNNSA-N | |
| 5369175 | |
| COC(=O)\C=C\CBr |
| C5H7BrO2 | |
| MFCD00000246,MFCD09998308 | |
| methyl 4-bromocrotonate, e-methyl 4-bromobut-2-enoate, 4-bromocrotonic acid methyl ester, methyl gamma-bromocrotonate, methyl bromocrotonate, methyl 4-bromo-2-butenoate, 2-butenoic acid, 4-bromo-, methyl ester, methyl 4-bromobut-2-enoate, methyl 2e-4-bromobut-2-enoate, crotonic acid, 4-bromo-, methyl ester | |
| methyl (2E)-4-bromobut-2-enoate |
Safety and Handling
RTECSNumber : GQ3120000
TSCA : Yes
Recommended Storage : Refrigerator