accessibility menu, dialog, popup

UserName

CAS RN 1117-71-1

IUPAC Name: methyl (2E)-4-bromobut-2-enoate
Synonyms: methyl (2E)-4-bromobut-2-enoate
Molecular Weight (g/mol): 179.01
Molecular Formula: C5H7BrO2
InChi Key: RWIKCBHOVNDESJ-NSCUHMNNSA-N
SMILES: COC(=O)\C=C\CBr
Boiling Point 
  • (2)
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (6)
  • (2)
  • (3)
  • (3)
  • (2)
  • (6)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (6)
  • (5)