missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Bromo-3-nitrobenzoate 98.0+%, TCI America™

Catalog No. M25961G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25961G 1 g
M25965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25961G Supplier TCI America Supplier No. M25961G

Chemical Identifiers

CAS 2363-16-8
Molecular Formula C8H6BrNO4
Molecular Weight (g/mol) 260.04
MDL Number MFCD00553089
InChI Key BNNDHGPPQZVKMX-UHFFFAOYSA-N
Synonym 4-Bromo-3-nitrobenzoic Acid Methyl Ester
PubChem CID 2736829
IUPAC Name methyl 4-bromo-3-nitrobenzoate
SMILES COC(=O)C1=CC=C(Br)C(=C1)[N+]([O-])=O

Specifications

Melting Point 103°C
Color White-Yellow
Quantity 1 g
Formula Weight 260.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Bromo-3-nitrobenzoate