accessibility menu, dialog, popup

UserName

CAS RN 2363-16-8

IUPAC Name: methyl 4-bromo-3-nitrobenzoate
Synonyms: methyl 4-bromo-3-nitrobenzoate
Molecular Weight (g/mol): 260.04
Molecular Formula: C8H6BrNO4
InChi Key: BNNDHGPPQZVKMX-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(Br)C(=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results