missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Bromo-3-methylbenzoate 97.0+%, TCI America™

Catalog No. p-7119583
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22565G 5 g
B225625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B22565G Supplier TCI America Supplier No. B22565G
Only null left

Chemical Identifiers

CAS 148547-19-7
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD00673014
InChI Key GTZTYNPAPQKIIR-UHFFFAOYSA-N
Synonym 4-bromo-3-methylbenzoic acid methyl ester, methyl4-bromo-3-methylbenzoate, methyl 4-bromo-m-toluate, 4-bromo-3-methyl-benzoic acid methyl ester, methyl 4-bromo-3-methyl-benzoate, 4-bromo-m-toluic acid methyl ester, benzoic acid, 4-bromo-3-methyl-, methyl ester, 2-bromo-5-methoxycarbonyl toluene, 3-methyl-4-bromobenzoic acid methyl ester, pubchem3970
PubChem CID 3294148
IUPAC Name methyl 4-bromo-3-methylbenzoate
SMILES CC1=C(C=CC(=C1)C(=O)OC)Br

Specifications

Melting Point 43°C
Color White-Yellow
Quantity 5 g
Formula Weight 229.07
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Bromo-3-methylbenzoate