accessibility menu, dialog, popup

UserName

CAS RN 148547-19-7

IUPAC Name: methyl 4-bromo-3-methylbenzoate
Synonyms: methyl 4-bromo-3-methylbenzoate
Molecular Weight (g/mol): 229.07
Molecular Formula: C9H9BrO2
InChi Key: GTZTYNPAPQKIIR-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(Br)C(C)=C1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)