missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-bromo-3-methylbenzoate, 97%

Catalog No. AAA1907806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1907806 5 g
AAA1907814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1907806 Supplier Thermo Scientific Chemicals Supplier No. A1907806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 148547-19-7 | C9H9BrO2 | 229.073 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 148547-19-7
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD00673014
InChI Key GTZTYNPAPQKIIR-UHFFFAOYSA-N
Synonym 4-bromo-3-methylbenzoic acid methyl ester, methyl4-bromo-3-methylbenzoate, methyl 4-bromo-m-toluate, 4-bromo-3-methyl-benzoic acid methyl ester, methyl 4-bromo-3-methyl-benzoate, 4-bromo-m-toluic acid methyl ester, benzoic acid, 4-bromo-3-methyl-, methyl ester, 2-bromo-5-methoxycarbonyl toluene, 3-methyl-4-bromobenzoic acid methyl ester, pubchem3970
PubChem CID 3294148
IUPAC Name methyl 4-bromo-3-methylbenzoate
SMILES CC1=C(C=CC(=C1)C(=O)OC)Br

Specifications

Melting Point 39°C to 44°C
Quantity 5 g
Beilstein 6593534
Formula Weight 229.07
Percent Purity 97%
Chemical Name or Material Methyl 4-bromo-3-methylbenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only