missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-bromo-2-fluorophenylacetate, 96%

Catalog No. p-7054348
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62482MD 250 mg
AAH6248203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH62482MD Supplier Thermo Scientific Chemicals Supplier No. H62482MD
Only null left

CAS: 193290-19-6 | C9H8BrFO2 | 247.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 193290-19-6
Molecular Formula C9H8BrFO2
Molecular Weight (g/mol) 247.06
MDL Number MFCD20483545
InChI Key QBGQCRUYARHQGO-UHFFFAOYSA-N
Synonym methyl 2-4-bromo-2-fluorophenyl acetate, methyl 4-bromo-2-fluorophenylacetate, methyl 4-bromo-2-fluorophenyl acetate, methyl 2-4-bromo-2-fluoro-phenyl acetate, 4-bromo-2-fluorophenyl acetic acid methyl ester, 4-bromo-2-fluoro-phenyl-acetic acid methyl ester, benzeneacetic acid, 4-bromo-2-fluoro-, methyl ester
PubChem CID 53429406
IUPAC Name methyl 2-(4-bromo-2-fluorophenyl)acetate
SMILES COC(=O)CC1=C(F)C=C(Br)C=C1

Specifications

Quantity 250 mg
Formula Weight 247.06
Percent Purity 96%
Physical Form Solid
Chemical Name or Material Methyl 4-bromo-2-fluorophenylacetate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only