CAS RN 193290-19-6
IUPAC Name:
methyl 2-(4-bromo-2-fluorophenyl)acetate
Synonyms:
methyl 2-(4-bromo-2-fluorophenyl)acetate
Molecular Weight (g/mol):
247.06
Molecular Formula:
C9H8BrFO2
InChi Key:
QBGQCRUYARHQGO-UHFFFAOYSA-N
SMILES:
COC(=O)CC1=C(F)C=C(Br)C=C1
1
–
1
of
1
results
1
Promotions Available
Methyl 4-bromo-2-fluorophenylacetate, 96%
Methyl 4-bromo-2-fluorophenylacetate, 96%
CAS: 193290-19-6 Molecular Formula: C9H8BrFO2 Molecular Weight (g/mol): 247.06 MDL Number: MFCD20483545 InChI Key: QBGQCRUYARHQGO-UHFFFAOYSA-N Synonym: methyl 2-4-bromo-2-fluorophenyl acetate,methyl 4-bromo-2-fluorophenylacetate,methyl 4-bromo-2-fluorophenyl acetate,methyl 2-4-bromo-2-fluoro-phenyl acetate,4-bromo-2-fluorophenyl acetic acid methyl ester,4-bromo-2-fluoro-phenyl-acetic acid methyl ester,benzeneacetic acid, 4-bromo-2-fluoro-, methyl ester PubChem CID: 53429406 IUPAC Name: methyl 2-(4-bromo-2-fluorophenyl)acetate SMILES: COC(=O)CC1=C(F)C=C(Br)C=C1