missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 4-Amino-2-methoxybenzoate 98.0+%, TCI America™
Supplier: TCI America M199825G
Specifications
| Methyl 4-Amino-2-methoxybenzoate | |
| 159°C | |
| C9H11NO3 | |
| 25 g | |
| YUPQMVSYNJQULF-UHFFFAOYSA-N | |
| methyl 4-amino-2-methoxybenzoate | |
| 168705 | |
| ≥98.0% (GC,T) |
| 27492-84-8 | |
| White-Yellow | |
| MFCD00017202 | |
| methyl 4-amino-o-anisate, 4-amino-2-methoxybenzoic acid methyl ester, benzoic acid, 4-amino-2-methoxy-, methyl ester, methyl 2-methoxy-4-aminobenzoate, methyl 4-amino-2-methoxy-benzoate, 4-amino-o-anisic acid methyl ester, methyl 4-amino-2-methoxybenzenecarboxylate, 2-methoxy-4-aminobenzoic acid methyl ester, ethylbenzyl dimethyltetradecylammonium chloride, pubchem4712 | |
| COC(=O)C1=CC=C(N)C=C1OC | |
| 181.19 | |
| 181.19 | |
| Crystalline Powder |
Chemical Identifiers
| 27492-84-8 | |
| 181.19 | |
| YUPQMVSYNJQULF-UHFFFAOYSA-N | |
| 168705 | |
| COC(=O)C1=CC=C(N)C=C1OC |
| C9H11NO3 | |
| MFCD00017202 | |
| methyl 4-amino-o-anisate, 4-amino-2-methoxybenzoic acid methyl ester, benzoic acid, 4-amino-2-methoxy-, methyl ester, methyl 2-methoxy-4-aminobenzoate, methyl 4-amino-2-methoxy-benzoate, 4-amino-o-anisic acid methyl ester, methyl 4-amino-2-methoxybenzenecarboxylate, 2-methoxy-4-aminobenzoic acid methyl ester, ethylbenzyl dimethyltetradecylammonium chloride, pubchem4712 | |
| methyl 4-amino-2-methoxybenzoate |
Safety and Handling
EINECSNumber : (9)-0102
TSCA : No