missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Amino-2-methoxybenzoate 98.0+%, TCI America™

Catalog No. p-7132957
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19985G 5 g
M199825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M19985G Supplier TCI America Supplier No. M19985G
Only null left

Chemical Identifiers

CAS 27492-84-8
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.19
MDL Number MFCD00017202
InChI Key YUPQMVSYNJQULF-UHFFFAOYSA-N
Synonym methyl 4-amino-o-anisate, 4-amino-2-methoxybenzoic acid methyl ester, benzoic acid, 4-amino-2-methoxy-, methyl ester, methyl 2-methoxy-4-aminobenzoate, methyl 4-amino-2-methoxy-benzoate, 4-amino-o-anisic acid methyl ester, methyl 4-amino-2-methoxybenzenecarboxylate, 2-methoxy-4-aminobenzoic acid methyl ester, ethylbenzyl dimethyltetradecylammonium chloride, pubchem4712
PubChem CID 168705
IUPAC Name methyl 4-amino-2-methoxybenzoate
SMILES COC(=O)C1=CC=C(N)C=C1OC

Specifications

Melting Point 159°C
Color White-Yellow
Quantity 5 g
Formula Weight 181.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Amino-2-methoxybenzoate
EINECSNumber (9)-0102
TSCA No