missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-Acetamido-2-methoxybenzoate 98.0+%, TCI America™

Catalog No. p-7133419
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27765G 5 g
M277625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27765G Supplier TCI America Supplier No. M27765G

Chemical Identifiers

CAS 4093-29-2
Molecular Formula C11H13NO4
Molecular Weight (g/mol) 223.23
MDL Number MFCD00065258
InChI Key OERVVBDWGVOBIS-UHFFFAOYSA-N
Synonym 4-Acetamido-2-methoxybenzoic Acid Methyl Ester, Methyl 4-Acetamido-o-anisate, 4-Acetamido-o-anisic Acid Methyl Ester
PubChem CID 77720
IUPAC Name methyl 4-acetamido-2-methoxybenzoate
SMILES COC(=O)C1=CC=C(NC(C)=O)C=C1OC

Specifications

Melting Point 130°C
Color White
Quantity 5 g
Formula Weight 223.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 4-Acetamido-2-methoxybenzoate
EINECSNumber (3)-1662
TSCA No