accessibility menu, dialog, popup

UserName

CAS RN 4093-29-2

IUPAC Name: methyl 4-acetamido-2-methoxybenzoate
Synonyms: methyl 4-acetamido-2-methoxybenzoate
Molecular Weight (g/mol): 223.23
Molecular Formula: C11H13NO4
InChi Key: OERVVBDWGVOBIS-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(NC(C)=O)C=C1OC
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)