missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4,5-dimethoxy-2-nitrobenzoate, 97%

Catalog No. AAA1283636
Change view
Click to view available options
Quantity:
50 g
250 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1283636 500 g
AAA1283618 50 g
AAA1283630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1283636 Supplier Thermo Scientific Chemicals Supplier No. A1283636
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 26791-93-5 | C10H11NO6 | 241.199 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 26791-93-5
Molecular Formula C10H11NO6
Molecular Weight (g/mol) 241.199
MDL Number MFCD00007239
InChI Key SYYKLKHBZGFKOC-UHFFFAOYSA-N
Synonym methyl 6-nitroveratrate, benzoic acid, 4,5-dimethoxy-2-nitro-, methyl ester, 6-nitroveratric acid methyl ester, 4,5-dimethoxy-2-nitrobenzoic acid, methyl ester, methyl 4,5-dimethoxy-2-nitro-benzoate, acmc-1cptq, ksc496m9r, methyl 3,4-dimethoxy-6-nitrobenzoate, methyl 3,4-dimethoxy-6-nitro-benzoate, methyl 4,5-dimethoxy-2-nitrobenzoate
PubChem CID 117867
IUPAC Name methyl 4,5-dimethoxy-2-nitrobenzoate
SMILES COC1=C(C=C(C(=C1)C(=O)OC)[N+](=O)[O-])OC

Specifications

Melting Point 142°C to 144°C
Quantity 500 g
Beilstein 2381747
Formula Weight 241.2
Percent Purity 97%
Chemical Name or Material Methyl 4,5-dimethoxy-2-nitrobenzoate
EINECSNumber 248-008-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only