accessibility menu, dialog, popup

UserName

CAS RN 26791-93-5

IUPAC Name: methyl 4,5-dimethoxy-2-nitrobenzoate
Synonyms: methyl 4,5-dimethoxy-2-nitrobenzoate
Molecular Weight (g/mol): 241.2
Molecular Formula: C10H11NO6
InChi Key: SYYKLKHBZGFKOC-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(OC)=C(OC)C=C1[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results