missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 4,4-Dimethyl-3-oxovalerate 95.0+%, TCI America™
$201.32 - $1,403.63
Chemical Identifiers
| CAS | 55107-14-7 |
|---|---|
| Molecular Formula | C8H14O3 |
| Molecular Weight (g/mol) | 158.197 |
| MDL Number | MFCD00008847 |
| InChI Key | XTXCFTMJPRXBBC-UHFFFAOYSA-N |
| Synonym | methyl pivaloylacetate, methyl 4,4-dimethyl-3-oxovalerate, methyl 4,4,4-trimethylacetoacetate, pentanoic acid, 4,4-dimethyl-3-oxo-, methyl ester, pivaloylacetic acid methyl ester, unii-99cq34ta4y, methyl 4,4,4-trimethyl-3-oxobutanoate, 4,4-dimethyl-3-oxovaleric acid methyl ester, benzyl s---lactate, acmc-209ll2 |
| PubChem CID | 99597 |
| IUPAC Name | methyl 4,4-dimethyl-3-oxopentanoate |
| SMILES | CC(C)(C)C(=O)CC(=O)OC |
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
|
P103125ML
|
TCI America
P103125ML |
25 mL |
N/A
|
|
|||||
|
P1031100ML
|
TCI America
P1031100ML |
100 mL |
N/A
|
|
|||||
|
P1031500ML
|
TCI America
P1031500ML |
500 mL |
N/A
|
|
|||||
Chemical Identifiers
| 55107-14-7 | |
| 158.197 | |
| XTXCFTMJPRXBBC-UHFFFAOYSA-N | |
| 99597 | |
| CC(C)(C)C(=O)CC(=O)OC |
| C8H14O3 | |
| MFCD00008847 | |
| methyl pivaloylacetate, methyl 4,4-dimethyl-3-oxovalerate, methyl 4,4,4-trimethylacetoacetate, pentanoic acid, 4,4-dimethyl-3-oxo-, methyl ester, pivaloylacetic acid methyl ester, unii-99cq34ta4y, methyl 4,4,4-trimethyl-3-oxobutanoate, 4,4-dimethyl-3-oxovaleric acid methyl ester, benzyl s---lactate, acmc-209ll2 | |
| methyl 4,4-dimethyl-3-oxopentanoate |
Specifications
| 55107-14-7 | |
| 84°C | |
| MFCD00008847 | |
| methyl pivaloylacetate, methyl 4,4-dimethyl-3-oxovalerate, methyl 4,4,4-trimethylacetoacetate, pentanoic acid, 4,4-dimethyl-3-oxo-, methyl ester, pivaloylacetic acid methyl ester, unii-99cq34ta4y, methyl 4,4,4-trimethyl-3-oxobutanoate, 4,4-dimethyl-3-oxovaleric acid methyl ester, benzyl s---lactate, acmc-209ll2 | |
| CC(C)(C)C(=O)CC(=O)OC | |
| 158.197 | |
| 158.20 | |
| Liquid |
| Yellow | |
| C8H14O3 | |
| 25 mL | |
| XTXCFTMJPRXBBC-UHFFFAOYSA-N | |
| methyl 4,4-dimethyl-3-oxopentanoate | |
| 99597 | |
| ≥95.0% (GC) | |
| Methyl 4,4-Dimethyl-3-oxovalerate |
Safety and Handling
EINECSNumber : (2)-1505
TSCA : Yes