accessibility menu, dialog, popup

UserName

CH3OOOCH3CH3H3C

CAS RN 55107-14-7

IUPAC Name: methyl 4,4-dimethyl-3-oxopentanoate
Synonyms: methyl 4,4-dimethyl-3-oxopentanoate
Molecular Weight (g/mol): 158.2
Molecular Formula: C8H14O3
InChi Key: XTXCFTMJPRXBBC-UHFFFAOYSA-N
SMILES: COC(=O)CC(=O)C(C)(C)C
Boiling Point 
  • (2)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (2)
  • (2)
  • (3)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)